missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid

Catalog No. AAJ64884MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64884MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64884MA Supplier Thermo Scientific Chemicals Supplier No. J64884MA
Only null left

CAS: 74341-63-2 | C7H10N2O4 | 186.167 g/mol

Selective quisqualate agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74341-63-2
Molecular Formula C7H10N2O4
Molecular Weight (g/mol) 186.167
MDL Number MFCD00213388
InChI Key UUDAMDVQRQNNHZ-UHFFFAOYSA-N
Synonym ampa, r,s-ampa, ampa pharmaceutical, alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, rs-ampa, +/--ampa, chembl13378, d,l-alpha-amino-3-hydroxy-5-methylisoxazole-4-propionic acid, r,s-alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, 4-isoxazolepropanoic acid, alpha-amino-2,3-dihydro-5-methyl-3-oxo
PubChem CID 1221
ChEBI CHEBI:34018
IUPAC Name 2-amino-3-(5-methyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
SMILES CC1=C(C(=O)NO1)CC(C(=O)O)N

Specifications

Color White
Quantity 10 mg
Solubility Information Soluble in warm water at 1mg/ml; Also soluble in 0;1M NaOH or 0;1M HCl at approximately 4mg/ml; Soluble in DMSO at 4mg/ml
Formula Weight 186.17
Physical Form Powder
Chemical Name or Material (+/-)-alpha-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only