missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha-Dimethyl-gamma-butyrolactone 97.0+%, TCI America™

Catalog No. D27335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27335G 5 g
D273325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27335G Supplier TCI America Supplier No. D27335G
Only null left

Chemical Identifiers

CAS 3709-08-8
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD03844788
InChI Key UPVAIJPDWVTFKT-UHFFFAOYSA-N
Synonym Dihydro-3,3-dimethyl-2(3H)-furanone
PubChem CID 94746
IUPAC Name 3,3-dimethyloxolan-2-one
SMILES CC1(CCOC1=O)C

Specifications

Melting Point 6°C
Color Yellow
Boiling Point 194°C
Quantity 5 g
Formula Weight 114.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha,alpha-Dimethyl-gamma-butyrolactone