Learn More
alpha,alpha'-Dibromo-m-xylene, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 626-15-3 |
|---|---|
| Molecular Formula | C8H8Br2 |
| Molecular Weight (g/mol) | 263.95 |
| InChI Key | OXHOPZLBSSTTBU-UHFFFAOYSA-N |
| Synonym | 1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene |
| PubChem CID | 69373 |
| IUPAC Name | 1,3-bis(bromomethyl)benzene |
| SMILES | C1=CC(=CC(=C1)CBr)CBr |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC155050050
|
Thermo Scientific Chemicals
155050050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC155050250
|
Thermo Scientific Chemicals
155050250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 626-15-3 | |
| 263.95 | |
| 1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene | |
| 1,3-bis(bromomethyl)benzene |
| C8H8Br2 | |
| OXHOPZLBSSTTBU-UHFFFAOYSA-N | |
| 69373 | |
| C1=CC(=CC(=C1)CBr)CBr |
Specifications
| 626-15-3 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| C6H4(CH2Br)2 | |
| 05,374 | |
| Solubility in water: insoluble | |
| C1=CC(=CC(=C1)CBr)CBr | |
| 263.95 | |
| 263.95 | |
| α, α'-Dibromo-m-xylene |
| 96.0 | |
| 135.0°C to 140.0°C (20.0mmHg) | |
| 96% min. (HPLC) | |
| C8H8Br2 | |
| 5g | |
| 1,3-bis bromomethyl benzene, m-xylylene dibromide, alpha,alpha'-dibromo-m-xylene, benzene, 1,3-bis bromomethyl, m-bis bromomethyl benzene, m-xylylene bromide, ccris 1777, m-xylyene dibromide, m-xylene, .alpha.,.alpha.'-dibromo, m-.alpha.,.alpha.'-dibromoxylene | |
| OXHOPZLBSSTTBU-UHFFFAOYSA-N | |
| 1,3-bis(bromomethyl)benzene | |
| 69373 | |
| 97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 210-931-6
RUO â Research Use Only