missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha'-Dianilino-p-xylene 98.0+%, TCI America™

Catalog No. p-7127132
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21431G 1 g
D21435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21431G Supplier TCI America Supplier No. D21431G
Only null left

Chemical Identifiers

CAS 13170-62-2
Molecular Formula C20H20N2
Molecular Weight (g/mol) 288.394
MDL Number MFCD00143578
InChI Key DXWQPWMYKQYRDS-UHFFFAOYSA-N
PubChem CID 607368
IUPAC Name N-[[4-(anilinomethyl)phenyl]methyl]aniline
SMILES C1=CC=C(C=C1)NCC2=CC=C(C=C2)CNC3=CC=CC=C3

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 1 g
Formula Weight 288.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha′-Dianilino-p-xylene