missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha',alpha'-Tetramethyl-1,3-benzenedipropionic Acid 98.0+%, TCI America™

Catalog No. T34201G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T34201G Supplier TCI America Supplier No. T34201G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 819050-88-9
Molecular Formula C16H22O4
Molecular Weight (g/mol) 278.35
MDL Number MFCD08276874
InChI Key BZWQVGTVYGEXTE-UHFFFAOYSA-N
Synonym 3,3′-(1,3-Phenylene)bis(2,2-dimethylpropanoic Acid)
PubChem CID 11204982
IUPAC Name 3-[3-(2-carboxy-2,2-dimethylethyl)phenyl]-2,2-dimethylpropanoic acid
SMILES CC(C)(CC1=CC(CC(C)(C)C(O)=O)=CC=C1)C(O)=O

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 1 g
Formula Weight 278.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha,alpha,alpha′,alpha′-Tetramethyl-1,3-benzenedipropionic Acid