missing translation for 'onlineSavingsMsg'
Learn More

alpha-Acetyl-gamma-butyrolactone 99.0+%, TCI America™

Catalog No. A068125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A068125G 25 g
A0681500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A068125G Supplier TCI America Supplier No. A068125G

Chemical Identifiers

CAS 517-23-7
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00005394
InChI Key OMQHDIHZSDEIFH-UHFFFAOYNA-N
Synonym 2-acetylbutyrolactone, 3-acetyldihydrofuran-2 3h-one, alpha-acetylbutyrolactone, alpha-acetyl-gamma-butyrolactone, 3-acetyldihydro-2 3h-furanone, 2 3h-furanone, 3-acetyldihydro, 3-acetyltetrahydrofuran-2-one, 2-oxo-3-acetyltetrahydrofuran, 3-acetyltetrahydro-2-furanone, 3-acetyl-2 3h-4,5-dihydrofuranone
PubChem CID 10601
IUPAC Name 3-acetyloxolan-2-one
SMILES CC(=O)C1CCOC1=O

Specifications

Color Colorless
Boiling Point 108°C
Quantity 25 g
Formula Weight 128.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Acetyl-gamma-butyrolactone
EINECSNumber (5)-0066
TSCA Yes