missing translation for 'onlineSavingsMsg'
Learn More

alpha-[2-(Methylamino)ethyl]benzyl Alcohol 98.0+%, TCI America™

Catalog No. M19605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19605G 5 g
M196025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19605G Supplier TCI America Supplier No. M19605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42142-52-9
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.236
MDL Number MFCD00674078
InChI Key XXSDCGNHLFVSET-UHFFFAOYSA-N
Synonym 3-Hydroxy-N-methyl-3-phenylpropylamine, 3-(Methylamino)-1-phenylpropanol
PubChem CID 2733989
IUPAC Name 3-(methylamino)-1-phenylpropan-1-ol
SMILES CNCCC(C1=CC=CC=C1)O

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 170°C
Quantity 5 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-[2-(Methylamino)ethyl]benzyl Alcohol