missing translation for 'onlineSavingsMsg'
Learn More

Aloe Vera, 200:1, Powder, Spectrum™ Chemical

A1861, 85507-69-3

$310.00 - $71263.51

Chemical Identifiers

CAS 85507-69-3
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18600427
View Documents
Spectrum Chemical Mfg Cor
A186125GM
25 g Amber Glass Bottle
Each for $310.00
Only null left
 
18600428
View Documents
Spectrum Chemical Mfg Cor
A1861100GM
100 g Amber Glass Bottle
Each for $858.97
Only null left
 
18600429
View Documents
Spectrum Chemical Mfg Cor
A1861500GM
500 g Amber Glass Bottle
Each for $2,372.66
Only null left
 
18600431
View Documents
Spectrum Chemical Mfg Cor
A186112KGBL
12 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18600430
View Documents
Spectrum Chemical Mfg Cor
A186150KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Aloe Vera, 200:1, Powder - Spectrum’s freeze dried aloe vera powder is easy to use. Just add 199 parts distilled or deionized water to 1 part aloe vera powder to reconstitute the oil derived from the Aloe Vera plant. It is known for its moisturizing and healing qualities and can be used in a wide range of personal care products. It is also a humectant, hair conditioners, and to relieve dandruff and dry scalp. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications

Chemical Identifiers

85507-69-3

Specifications

85507-69-3
Amber Glass Bottle
Reagent
100%
25 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'