missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Alnico Bar Magnets

Catalog No. S15862
Change view
Click to view available options
Dimensions (L x W x H):
1.5 x 0.5 x .25 in.
2 x 0.5 x .25 in.
3 x 0.5 x .25 in.
4 x 0.5 x .25 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S15862 3 x 0.5 x .25 in.
S15705 1.5 x 0.5 x .25 in.
S15704 2 x 0.5 x .25 in.
S15773 4 x 0.5 x .25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S15862 Supplier United Scientific™ Supplier No. ABM030

These Alnico Bar Magnets are powerful magnets for classroom experiments.

  • Powerful Alnico 1 bar magnets, shipped in pairs, with end keepers.
  • Magnets are painted with North and South poles, clearly marked.
  • Magnets available in five different sizes.

Specifications

Material Alnico
For Use With (Application) Magnetic Demonstrations
Grade 5+
Product Type Magnet
Dimensions (L x W x H) 3 x 0.5 x .25 in.

Warning: Cancer and reproductive harm