missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Alnico Bar Magnets with Spacer and Keepers

Catalog No. S14041
Change view
Click to view available options
Grade:
9 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S14041 9 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S14041 Supplier Eisco™ Supplier No. PH0789D

Alnico Bar Magnets with Spacer and Keepers supplied in cardboard boxes.

  • With spacer and keepers
  • Supplied in cardboard boxes

Specifications

Description 1 set
Shape Bar
Grade 9 to 12
Color Red
Product Type Alnico Bar Magnet
Includes One pair
Dimensions (L x W x H) 150 x 12 x 6 mm
Quantity 1