missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Allylurea | |
| 88°C | |
| C4H8N2O | |
| 25 g | |
| VPJDULFXCAQHRC-UHFFFAOYSA-N | |
| prop-2-enylurea | |
| 11181 | |
| ≥98.0% (N) |
| 557-11-9 | |
| White | |
| MFCD00007954 | |
| allylurea, 1-allylurea, n-allylurea, urea, 2-propenyl, allylcarbamide, monoallylurea, allyl urea, n-2-propenylurea, urea, allyl, 2-propenylurea | |
| C=CCNC(=O)N | |
| 100.121 | |
| 100.12 | |
| Crystalline Powder |
Chemical Identifiers
| 557-11-9 | |
| 100.121 | |
| VPJDULFXCAQHRC-UHFFFAOYSA-N | |
| 11181 | |
| C=CCNC(=O)N |
| C4H8N2O | |
| MFCD00007954 | |
| allylurea, 1-allylurea, n-allylurea, urea, 2-propenyl, allylcarbamide, monoallylurea, allyl urea, n-2-propenylurea, urea, allyl, 2-propenylurea | |
| prop-2-enylurea |
Safety and Handling
RTECSNumber : YR7875000
TSCA : Yes