missing translation for 'onlineSavingsMsg'
Learn More

Allyltris(trimethylsilyloxy)silane 96.0+%, TCI America™

Catalog No. a21111g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21111G 1 g
A21115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21111G Supplier TCI America Supplier No. A21111G
Only null left

Chemical Identifiers

CAS 7087-21-0
Molecular Formula C12H32O3Si4
Molecular Weight (g/mol) 336.725
MDL Number MFCD00054804
InChI Key WCAXVXQTTCIZHD-UHFFFAOYSA-N
PubChem CID 21902346
IUPAC Name trimethyl-[prop-2-enyl-bis(trimethylsilyloxy)silyl]oxysilane
SMILES C[Si](C)(C)O[Si](CC=C)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 111°C
Quantity 1 g
Formula Weight 336.73
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyltris(trimethylsilyloxy)silane