missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyltrichlorosilane, 95%
Description
Allyltrichlorosilane acts as a reagent that produces homoallylic alcohols upon reaction with aldehydes in DMF. It is used to make silicones and glass fiber fininshes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Allyltrichlorosilane acts as a reagent that produces homoallylic alcohols upon reaction with aldehydes in DMF. It is used to make silicones and glass fiber fininshes.
Solubility
Soluble in THF, toluene, benzene. Reacts with water.
Notes
Moisture Sensitive. Store at 4°C. Store away from oxidizing agents and moisture. Incompatible with water.
Allyltrichlorosilane acts as a reagent that produces homoallylic alcohols upon reaction with aldehydes in DMF. It is used to make silicones and glass fiber fininshes.
Solubility
Soluble in THF, toluene, benzene. Reacts with water.
Notes
Moisture Sensitive. Store at 4°C. Store away from oxidizing agents and moisture. Incompatible with water.
Specifications
Specifications
| Boiling Point | 117°C to 118°C |
| Quantity | 5 g |
| Linear Formula | CH2=CHCH2SiCl3 |
| UN Number | UN1724 |
| Beilstein | 1743449 |
| Flash Point | 18°C (64°F) |
| Solubility Information | Soluble in THF,toluene,benzene. Reacts with water. |
| Formula Weight | 175.52 |
| Percent Purity | 95% |
| Refractive Index | 1.449 |
| Show More |
Safety and Handling
| Hazard Category | H225-H314 |
| Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: II |
| EINECSNumber | 203-485-9 |
| RTECSNumber | VV1530000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only