missing translation for 'onlineSavingsMsg'
Learn More

Allyltrichlorosilane, 95%

Catalog No. AAL0370306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0370306 5 g
AAL0370314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0370306 Supplier Thermo Scientific Chemicals Supplier No. L0370306
Only null left

CAS: 107-37-9 | C3H5Cl3Si | 175.508 g/mol

Allyltrichlorosilane acts as a reagent that produces homoallylic alcohols upon reaction with aldehydes in DMF. It is used to make silicones and glass fiber fininshes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltrichlorosilane acts as a reagent that produces homoallylic alcohols upon reaction with aldehydes in DMF. It is used to make silicones and glass fiber fininshes.

Solubility
Soluble in THF, toluene, benzene. Reacts with water.

Notes
Moisture Sensitive. Store at 4°C. Store away from oxidizing agents and moisture. Incompatible with water.

Chemical Identifiers

CAS 107-37-9
Molecular Formula C3H5Cl3Si
Molecular Weight (g/mol) 175.508
MDL Number MFCD00000486
InChI Key HKFSBKQQYCMCKO-UHFFFAOYSA-N
Synonym allyltrichlorosilane, silane, trichloro-2-propenyl, silane, allyltrichloro, silane, trichloroallyl, allyl trichlorosilane, propen-3-yltrichlorosilane, allytrichlorosilane, allylsilicone trichloride, unii-ub3n98803n, silane, trichloro-2-propen-1-yl
PubChem CID 7867
IUPAC Name trichloro(prop-2-enyl)silane
SMILES C=CC[Si](Cl)(Cl)Cl

Specifications

Boiling Point 117°C to 118°C
Quantity 5 g
Linear Formula CH2=CHCH2SiCl3
UN Number UN1724
Beilstein 1743449
Flash Point 18°C (64°F)
Solubility Information Soluble in THF,toluene,benzene. Reacts with water.
Formula Weight 175.52
Percent Purity 95%
Refractive Index 1.449
Sensitivity Moisture sensitive
Density 1.211
Chemical Name or Material Allyltrichlorosilane
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 203-485-9
RTECSNumber VV1530000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only