missing translation for 'onlineSavingsMsg'
Learn More

Allyltributyltin 95.0+%, TCI America™

Catalog No. p-7117314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12225G 5 g
A122225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12225G Supplier TCI America Supplier No. A12225G
Only null left

Chemical Identifiers

CAS 24850-33-7
Molecular Formula C15H32Sn
Molecular Weight (g/mol) 331.13
MDL Number MFCD00010346
InChI Key YLGRTLMDMVAFNI-UHFFFAOYSA-N
Synonym allyltributyltin, allyltributylstannane, allyltri-n-butyltin, stannane, tributyl-2-propenyl, tributyl prop-2-en-1-yl stannane, allyl tributyl stannane, unii-aor8od4103, tributyl-2-propenylstannane, tri-n-butyl-2-propenylstannane, allyl tributyltin
PubChem CID 90628
IUPAC Name tributyl(prop-2-en-1-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)CC=C

Specifications

Color Colorless
Boiling Point 155°C
Quantity 5 g
UN Number 2788
Formula Weight 331.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyltributyltin