missing translation for 'onlineSavingsMsg'
Learn More

Allyltributyltin, 97%

Catalog No. AC265550050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC265550050 5 g Glass bottle
AC265550250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC265550050 Supplier Thermo Scientific Chemicals Supplier No. 265550050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24850-33-7 | C15H32Sn | 331.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24850-33-7
Molecular Formula C15H32Sn
Molecular Weight (g/mol) 331.13
MDL Number MFCD00010346
InChI Key YLGRTLMDMVAFNI-UHFFFAOYSA-N
Synonym allyltributyltin, allyltributylstannane, allyltri-n-butyltin, stannane, tributyl-2-propenyl, tributyl prop-2-en-1-yl stannane, allyl tributyl stannane, unii-aor8od4103, tributyl-2-propenylstannane, tri-n-butyl-2-propenylstannane, allyl tributyltin
PubChem CID 90628
IUPAC Name tributyl(prop-2-enyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)CC=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.0680g/mL
Boiling Point 88.0°C to 92.0°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4850 to 1.4870
Linear Formula [CH3(CH2)3]3SnCH2CH=CH2
Quantity 5 g
Beilstein 04, 1922
Fieser 09,8; 11,15; 12,21; 14,14; 16,7; 17,12
Specific Gravity 1.068
Merck Index 15, 289
Formula Weight 331.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Allyltributyltin
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 246-494-3

RUO – Research Use Only