missing translation for 'onlineSavingsMsg'
Learn More

Allylcyclohexane 97.0+%, TCI America™

Numéro de catalogue. A12015ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A12015ML 5 mL
A120125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A12015ML Fournisseur TCI America Code fournisseur A12015ML
Il en reste null

Identifiants chimiques

CAS 2114-42-3
Molecular Formula C9H16
Molecular Weight (g/mol) 124.227
MDL Number MFCD00013769
InChI Key KVOZXXSUSRZIKD-UHFFFAOYSA-N
Synonym allylcyclohexane, allyl-cyclohexane, cyclohexane, allyl, 3-cyclohexyl-1-propene, cyclohexane, 2-propenyl, 1-cyclohexyl-2-propene, 1-propene, 3-cyclohexyl, allyl cyclohexane, 3-cyclohexylpropene, 2-propenyl-cyclohexane
PubChem CID 75027
IUPAC Name prop-2-enylcyclohexane
SMILES C=CCC1CCCCC1

Spécifications

Color Colorless
Boiling Point 151°C
Quantity 5 mL
UN Number 3295
Formula Weight 124.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allylcyclohexane