missing translation for 'onlineSavingsMsg'
Learn More

Allylchlorodimethylsilane 96.0+%, TCI America™

Catalog No. A127525ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A127525ML 25 mL
A127510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A127525ML Supplier TCI America Supplier No. A127525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4028-23-3
Molecular Formula C5H11ClSi
Molecular Weight (g/mol) 134.678
MDL Number MFCD00000506
InChI Key KMVZWUQHMJAWSY-UHFFFAOYSA-N
Synonym allyldimethylchlorosilane, allylchlorodimethylsilane, silane, chlorodimethyl-2-propenyl, allyl chloro dimethylsilane, allyl-chloro-dimethyl silane, silane, chlorodimethyl-2-propen-1-yl, admcs, allyl chlorodimethylsilane, allylchlorodimethyl silane, acmc-1afr7
PubChem CID 77646
IUPAC Name chloro-dimethyl-prop-2-enylsilane
SMILES C[Si](C)(CC=C)Cl

Specifications

Color Yellow
Boiling Point 113°C
Quantity 25 mL
UN Number 2985
Formula Weight 134.68
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Allylchlorodimethylsilane [Allyldimethylsilylating Reagent for GC/MS]
EINECSNumber (2)-2042
TSCA Yes
Recommended Storage Refrigerator