missing translation for 'onlineSavingsMsg'
Learn More

Allyl trifluoroacetate, 97%

Catalog No. p-7081641
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1169009 10 g
AAA1169018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1169009 Supplier Thermo Scientific Chemicals Supplier No. A1169009
Only null left

CAS: 383-67-5 | C5H5F3O2 | 154.088 g/mol

Used as nucleophilic ally metal precursor in alkylation reaction that generates homoallylic amines. Also used as chemical reagent and intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as nucleophilic ally metal precursor in alkylation reaction that generates homoallylic amines. Also used as chemical reagent and intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 383-67-5
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.088
MDL Number MFCD00013567
InChI Key XIVPVSIDXBTZLM-UHFFFAOYSA-N
Synonym allyl trifluoroacetate, allyl trifluoracetate, trifluoroacetic acid allyl ester, prop-2-en-1-yl 2,2,2-trifluoroacetate, allyltrifluoroacetate, pubchem12548, acmc-209izu, allyl alcohol, trifluoroacetate, acetic acid, trifluoro-, allyl ester
PubChem CID 67845
IUPAC Name prop-2-enyl 2,2,2-trifluoroacetate
SMILES C=CCOC(=O)C(F)(F)F

Specifications

Density 1.183
Boiling Point 73°C to 75°C
Flash Point −1°C (30°F)
Refractive Index 1.336
Quantity 10 g
UN Number UN2924
Beilstein 1766312
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 154.09
Percent Purity 97%
Chemical Name or Material Allyl trifluoroacetate
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 206-853-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only