missing translation for 'onlineSavingsMsg'
Learn More

Allyl n-Octyl Ether 95.0+%, TCI America™

Numéro de catalogue. A105210ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A105210ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A105210ML Fournisseur TCI America Code fournisseur A105210ML
Il en reste null

Identifiants chimiques

CAS 3295-97-4
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
MDL Number MFCD00015294
InChI Key IELYMBBIHQDONA-UHFFFAOYSA-N
PubChem CID 520600
IUPAC Name 1-prop-2-enoxyoctane
SMILES CCCCCCCCOCC=C

Spécifications

Color Colorless
Boiling Point 206°C
Quantity 10 mL
Formula Weight 170.30
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl n-Octyl Ether