missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl Methyl Carbonate 98.0+%, TCI America™
$86.64 - $346.52
Chemical Identifiers
| CAS | 35466-83-2 |
|---|---|
| Molecular Formula | C5H8O3 |
| Molecular Weight (g/mol) | 116.116 |
| MDL Number | MFCD00134545 |
| InChI Key | YHLVIDQQTOMBGN-UHFFFAOYSA-N |
| Synonym | allyl methyl carbonate, methyl prop-2-en-1-yl carbonate, carbonic acid allyl methyl ester, isobutylene carbonate, methyl allyl carbonate, acmc-1agon, allyl methyl carbonate #, prop-2-enyl methoxyformate, yhlvidqqtombgn-uhfffaoysa |
| PubChem CID | 534788 |
| IUPAC Name | methyl prop-2-enyl carbonate |
| SMILES | COC(=O)OCC=C |
Chemical Identifiers
| 35466-83-2 | |
| 116.116 | |
| YHLVIDQQTOMBGN-UHFFFAOYSA-N | |
| 534788 | |
| COC(=O)OCC=C |
| C5H8O3 | |
| MFCD00134545 | |
| allyl methyl carbonate, methyl prop-2-en-1-yl carbonate, carbonic acid allyl methyl ester, isobutylene carbonate, methyl allyl carbonate, acmc-1agon, allyl methyl carbonate #, prop-2-enyl methoxyformate, yhlvidqqtombgn-uhfffaoysa | |
| methyl prop-2-enyl carbonate |
Specifications
| 35466-83-2 | |
| 131°C | |
| MFCD00134545 | |
| 3272 | |
| YHLVIDQQTOMBGN-UHFFFAOYSA-N | |
| methyl prop-2-enyl carbonate | |
| 534788 | |
| ≥98.0% (GC) | |
| Allyl Methyl Carbonate |
| Colorless | |
| C5H8O3 | |
| 5 mL | |
| allyl methyl carbonate, methyl prop-2-en-1-yl carbonate, carbonic acid allyl methyl ester, isobutylene carbonate, methyl allyl carbonate, acmc-1agon, allyl methyl carbonate #, prop-2-enyl methoxyformate, yhlvidqqtombgn-uhfffaoysa | |
| COC(=O)OCC=C | |
| 116.116 | |
| 116.12 | |
| Liquid |
Safety and Handling
TSCA : No