missing translation for 'onlineSavingsMsg'
Learn More

Allyl Methyl Carbonate 98.0+%, TCI America™

Catalog No. C13155ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13155ML 5 mL
C131525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13155ML Supplier TCI America Supplier No. C13155ML

Chemical Identifiers

CAS 35466-83-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00134545
InChI Key YHLVIDQQTOMBGN-UHFFFAOYSA-N
Synonym allyl methyl carbonate, methyl prop-2-en-1-yl carbonate, carbonic acid allyl methyl ester, isobutylene carbonate, methyl allyl carbonate, acmc-1agon, allyl methyl carbonate #, prop-2-enyl methoxyformate, yhlvidqqtombgn-uhfffaoysa
PubChem CID 534788
IUPAC Name methyl prop-2-enyl carbonate
SMILES COC(=O)OCC=C

Specifications

Color Colorless
Boiling Point 131°C
Quantity 5 mL
UN Number 3272
Formula Weight 116.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Methyl Carbonate