missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl methyl carbonate, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Density | 1.0220g/mL |
| Boiling Point | 131.0°C |
| Flash Point | 33°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4050 to 1.4070 (20°C, 589nm) |
| Linear Formula | CH2=CHCH2OCO2CH3 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present a |
RUO – Research Use Only