missing translation for 'onlineSavingsMsg'
Learn More

Allyl Methacrylate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. p-7131970
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M007525ML 25 mL
M0075500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M007525ML Supplier TCI America Supplier No. M007525ML
Only null left

Chemical Identifiers

CAS 96-05-9
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00008592
InChI Key FBCQUCJYYPMKRO-UHFFFAOYSA-N
Synonym allyl methacrylate, ageflex ama, methacrylic acid, allyl ester, 2-propenoic acid, 2-methyl-, 2-propenyl ester, allylmethacrylate, allyl 2-methylacrylate, allylester kyseliny methakrylove, methacrylic acid allyl ester, unii-g2ig50653z, allylester kyseliny methakrylove czech
PubChem CID 7274
IUPAC Name prop-2-en-1-yl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC=C

Specifications

Color Colorless
Boiling Point 64°C
Quantity 25 mL
UN Number 2929
Formula Weight 126.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1037
RTECSNumber UD3483000
TSCA Yes