missing translation for 'onlineSavingsMsg'
Learn More

Allyl Hexanoate 98.0+%, TCI America™

Catalog No. A217925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A217925ML 25 mL
A2179500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A217925ML Supplier TCI America Supplier No. A217925ML
Only null left

Chemical Identifiers

CAS 123-68-2
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.225
MDL Number MFCD00038339
InChI Key RCSBILYQLVXLJG-UHFFFAOYSA-N
Synonym Hexanoic Acid Allyl Ester
PubChem CID 31266
IUPAC Name prop-2-enyl hexanoate
SMILES CCCCCC(=O)OCC=C

Specifications

Color Colorless
Boiling Point 188°C
Quantity 25 mL
UN Number 2810
Formula Weight 156.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Hexanoate
EINECSNumber (2)-0759
RTECSNumber MO6125000
TSCA Yes