missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl Heptanoate 98.0+%, TCI America™
$188.67 - $1137.77
Chemical Identifiers
| CAS | 142-19-8 |
|---|---|
| Molecular Formula | C10H18O2 |
| Molecular Weight (g/mol) | 170.252 |
| MDL Number | MFCD00038341 |
| InChI Key | SJWKGDGUQTWDRV-UHFFFAOYSA-N |
| Synonym | allyl heptanoate, allyl heptylate, 2-propenyl heptanoate, allyl enanthate, allyl heptoate, heptanoic acid, 2-propenyl ester, prop-2-en-1-yl heptanoate, heptanoic acid, allyl ester, allylester kyseliny enanthove, heptanoic acid, 2-propen-1-yl ester |
| PubChem CID | 8878 |
| IUPAC Name | prop-2-enyl heptanoate |
| SMILES | CCCCCCC(=O)OCC=C |
Chemical Identifiers
| 142-19-8 | |
| 170.252 | |
| SJWKGDGUQTWDRV-UHFFFAOYSA-N | |
| 8878 | |
| CCCCCCC(=O)OCC=C |
| C10H18O2 | |
| MFCD00038341 | |
| allyl heptanoate, allyl heptylate, 2-propenyl heptanoate, allyl enanthate, allyl heptoate, heptanoic acid, 2-propenyl ester, prop-2-en-1-yl heptanoate, heptanoic acid, allyl ester, allylester kyseliny enanthove, heptanoic acid, 2-propen-1-yl ester | |
| prop-2-enyl heptanoate |
Specifications
| 142-19-8 | |
| Yellow | |
| C10H18O2 | |
| 25 g | |
| allyl heptanoate, allyl heptylate, 2-propenyl heptanoate, allyl enanthate, allyl heptoate, heptanoic acid, 2-propenyl ester, prop-2-en-1-yl heptanoate, heptanoic acid, allyl ester, allylester kyseliny enanthove, heptanoic acid, 2-propen-1-yl ester | |
| CCCCCCC(=O)OCC=C | |
| 170.252 | |
| 170.25 | |
| Liquid |
| -66°C | |
| 89°C | |
| MFCD00038341 | |
| 2810 | |
| SJWKGDGUQTWDRV-UHFFFAOYSA-N | |
| prop-2-enyl heptanoate | |
| 8878 | |
| ≥98.0% (GC) | |
| Allyl Heptanoate |
Safety and Handling
EINECSNumber : (2)-0759
RTECSNumber : MJ1750000
TSCA : Yes