missing translation for 'onlineSavingsMsg'
Learn More

Allyl Glycidyl Ether 99.0+%, TCI America™

Catalog No. p-7116771
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
A022125ML 25 mL
A0221500ML 500 mL
2 options

Catalog No. A022125ML

Supplier: TCI America A022125ML

Only null left

Chemical Identifiers

CAS 106-92-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00005143
InChI Key LSWYGACWGAICNM-UHFFFAOYSA-N
Synonym allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
PubChem CID 7838
IUPAC Name 2-(prop-2-enoxymethyl)oxirane
SMILES C=CCOCC1CO1

Specifications

Melting Point -100°C
Color Colorless
Boiling Point 154°C
Quantity 25 mL
UN Number 2219
Formula Weight 114.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Glycidyl Ether
EINECSNumber (2)-0393
RTECSNumber RR0875000
TSCA Yes