missing translation for 'onlineSavingsMsg'
Learn More

Allyl Glycidyl Ether 99.0+%, TCI America™

Catalog No. A022125ML
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A022125ML 25 mL
A0221500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. A022125ML missing translation for 'mfr' TCI America missing translation for 'supplierNo' A022125ML

Chemical Identifiers

CAS 106-92-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00005143
InChI Key LSWYGACWGAICNM-UHFFFAOYSA-N
Synonym allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
PubChem CID 7838
IUPAC Name 2-(prop-2-enoxymethyl)oxirane
SMILES C=CCOCC1CO1

Specifications

Melting Point -100°C
Color Colorless
Boiling Point 154°C
Quantity 25 mL
UN Number 2219
Formula Weight 114.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Glycidyl Ether
EINECSNumber (2)-0393
RTECSNumber RR0875000
TSCA Yes