missing translation for 'onlineSavingsMsg'
Learn More

Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific™

Chemical Identifiers

CAS 106-92-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00005143
InChI Key LSWYGACWGAICNM-UHFFFAOYSA-N
Synonym allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
PubChem CID 7838
IUPAC Name 2-(prop-2-enoxymethyl)oxirane
SMILES C=CCOCC1CO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC430951000
SDS View Documents
Thermo Scientific Chemicals
430951000
100mL AcroSeal™ Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC430958000
SDS View Documents
Thermo Scientific Chemicals
430958000
800mL AcroSeal™ Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

106-92-3
114.14
LSWYGACWGAICNM-UHFFFAOYSA-N
7838
C=CCOCC1CO1
C6H10O2
MFCD00005143
allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
2-(prop-2-enoxymethyl)oxirane

Specifications

106-92-3
100.0
154.0°C
Authentic
AcroSeal™ Glass Bottle
1.4332 to 1.4352
100mL
allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
LSWYGACWGAICNM-UHFFFAOYSA-N
2-(prop-2-enoxymethyl)oxirane
7838
99+%
99.0
0.9620g/mL
48°C
99% min. (GC)
C6H10O2
MFCD00005143
0.962
Solubility in water: 50g/L (20°C). Other solubilities: soluble in ethanol,acetone,diethyl ether and,benzene
C=CCOCC1CO1
114.14
114.14
Allyl glycidyl ether
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Suspected of causing cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility.
Toxic if inhaled.
Harmful if swallowed.
May cause respiratory irri

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

EINECSNumber : 203-442-4

SDS
Documents

Documents

RUO – Research Use Only