missing translation for 'onlineSavingsMsg'
Learn More

Allyl glycidyl ether, 97%

Catalog No. p-7045699
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1163422 100 g
AAL1163436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1163422 Supplier Thermo Scientific Chemicals Supplier No. L1163422
Only null left

CAS: 106-92-3 | C6H10O2 | 114.144 g/mol

Allyl glycidyl ether is useful in hydrosilylation, including the H2PtCl6·xH2O (cat. no. 206083) catalyzed hydrosilylation of siloxanes to epoxysiloxanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl glycidyl ether is useful in hydrosilylation, including the H2PtCl6·xH2O (cat. no. 206083) catalyzed hydrosilylation of siloxanes to epoxysiloxanes.

Solubility
Soluble in water (140 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 106-92-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00005143
InChI Key LSWYGACWGAICNM-UHFFFAOYSA-N
Synonym allyl glycidyl ether, glycidyl allyl ether, allylglycidaether, oxirane, 2-propenyloxy methyl, allyl 2,3-epoxypropyl ether, allil-glicidil-etere, neoallyl g, 1,2-epoxy-3-allyloxypropane, 1-allyloxy-2,3-epoxypropane, santolin xi
PubChem CID 7838
IUPAC Name 2-(prop-2-enoxymethyl)oxirane
SMILES C=CCOCC1CO1

Specifications

Melting Point -100°C
Density 0.967
Boiling Point 50°C to 51°C (15 mmHg)
Flash Point 47°C (134°F)
Odor Ether-like
Refractive Index 1.435
Quantity 100 g
UN Number UN2219
Beilstein 105871
Solubility Information Soluble in water (140g/L at 20°C).
Formula Weight 114.15
Percent Purity 97%
Chemical Name or Material Allyl glycidyl ether
Show More Show Less
Hazard Category H226-H302-H315-H317-H318-H331-H335-H341-H351-H361f-H373
Hazard Statement GHS H Statement
H318-H226-H341-H351-H361-H302-H332-H317-H335-H315
Causes serious eye damage.
Flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P311-P330-P333+P313-P363-P37
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALLYL GLYCIDYL ETHER
EINECSNumber 203-442-4
RTECSNumber RR0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only