missing translation for 'onlineSavingsMsg'
Learn More

Allyl Ethyl Ether 95.0+%, TCI America™

Catalog No. p-7117202
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
A10335ML 5 mL
A103325ML 25 mL
2 options

Catalog No. A10335ML

Supplier: TCI America A10335ML

Only null left

Chemical Identifiers

CAS 557-31-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00009255
InChI Key OJPSFJLSZZTSDF-UHFFFAOYSA-N
Synonym allyl ethyl ether, 1-propene, 3-ethoxy, allylethylether, ethyl allyl ether, unii-sm8t658re3, ether, allyl ethyl, 3-ethoxy-1-propene, ethyl 2-propenyl ether, 3-ethoxypropene, acmc-1atoi
PubChem CID 11191
IUPAC Name 3-ethoxyprop-1-ene
SMILES CCOCC=C

Specifications

Color Yellow
Boiling Point 67°C
Quantity 5 mL
UN Number 2335
Formula Weight 86.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Ethyl Ether