missing translation for 'onlineSavingsMsg'
Learn More

Allyl Cyclohexanepropionate 98.0+%, TCI America™

Catalog No. C048525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C048525ML 25 mL
C0485500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C048525ML Supplier TCI America Supplier No. C048525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2705-87-5
Molecular Formula C12H20O2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00059557
InChI Key TWXUTZNBHUWMKJ-UHFFFAOYSA-N
Synonym Cyclohexanepropionic Acid Allyl Ester, 3-Cyclohexylpropionic Acid Allyl Ester
PubChem CID 17617
ChEBI CHEBI:31188
IUPAC Name prop-2-enyl 3-cyclohexylpropanoate
SMILES C=CCOC(=O)CCC1CCCCC1

Specifications

Color Colorless
Boiling Point 91°C
Quantity 25 mL
Formula Weight 196.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Cyclohexanepropionate
EINECSNumber (3)-2429
RTECSNumber GV6735000
TSCA Yes