missing translation for 'onlineSavingsMsg'
Learn More

Allyl chloroformate, 97%

Catalog No. AC221741000
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221741000 100 mL Glass bottle
AC221740500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC221741000 Supplier Thermo Scientific Chemicals Supplier No. 221741000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2937-50-0 | C4H5ClO2 | 120.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 2937-50-0
Molecular Formula C4H5ClO2
Molecular Weight (g/mol) 120.54
MDL Number MFCD00000648
InChI Key CAEWJEXPFKNBQL-UHFFFAOYSA-N
Synonym allyl chloroformate, chloroformic acid allyl ester, allyl chlorocarbonate, allylchloroformate, 2-propenyl chloroformate, carbonochloridic acid, 2-propenyl ester, formic acid, chloro-, allyl ester, hsdb 621, allylester kyseliny chlormravenci, allylester kyseliny chlormravenci czech
PubChem CID 18052
IUPAC Name prop-2-enyl carbonochloridate
SMILES C=CCOC(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown-Red to Yellow
Density 1.1300g/mL
Boiling Point 109.0°C to 110.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4213 to 1.4233
Linear Formula ClCO2CH2CH=CH2
Quantity 100 mL
Beilstein 03, 12
Fieser 12,15; 13,9
Specific Gravity 1.13
Solubility Information Solubility in water: decomposes
Formula Weight 120.54
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Allyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
EINECSNumber 220-916-6

RUO – Research Use Only