missing translation for 'onlineSavingsMsg'
Learn More

Allyl(chloro)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II), Thermo Scientific Chemicals

Catalog No. AA4674703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4674703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4674703 Supplier Thermo Scientific Chemicals Supplier No. 04674703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1248656-98-5
Molecular Formula C51H52ClFePPd
Molecular Weight (g/mol) 893.666
MDL Number MFCD25372546
InChI Key SURRVSCVVZPMFV-UHFFFAOYSA-M
Synonym pdclallyl qphos, allyl chloro 1,2,3,4,5-pentaphenyl-1'-di-tert-butylphosphino ferrocene palladium ii
PubChem CID 73994982
IUPAC Name chloropalladium(1+);ditert-butyl(cyclopenta-2,4-dien-1-yl)phosphane;iron(2+);prop-1-ene;(2,3,4,5-tetraphenylcyclopenta-1,4-dien-1-yl)benzene
SMILES CC(C)(C)P([C-]1C=CC=C1)C(C)(C)C.[CH2-]C=C.C1=CC=C(C=C1)[C-]2C(=C(C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6.Cl[Pd+].[Fe+2]

Specifications

Color Pink
Quantity 1 g
Solubility Information It is soluble in CH2Cl2 and insoluble in hexane.
Formula Weight 893.67
Physical Form Solid
Chemical Name or Material Allyl(chloro)[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(II)
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only