Learn More
Allyl Alcohol, 99%, extra pure
CAS: 107-18-6 | C3H6O | 58.08 g/mol
Supplier: Thermo Scientific Chemicals 102860010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Allyl alcohol | |
| 107-18-6 | |
| 100.0 | |
| 0.8500g/mL | |
| 21°C | |
| 98.5% min. (GC) | |
| C3H6O | |
| H2C=CHCH2OH | |
| 1 L | |
| 0.85 | |
| allyl alcohol, 2-propen-1-ol, vinylcarbinol, 2-propenyl alcohol, 3-hydroxypropene, 2-propenol, allylic alcohol, 1-propen-3-ol, vinyl carbinol, weed drench | |
| XXROGKLTLUQVRX-UHFFFAOYSA-N | |
| prop-2-en-1-ol | |
| 7858 | |
| 58.08 | |
| Extra Pure |
| Extra Pure | |
| 98.5 | |
| -129.0°C | |
| 96.0°C to 98.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4119 to 1.4139 | |
| MFCD00002920 | |
| 01,436 | |
| 15,277 | |
| Solubility in water: miscible. Other solubilities: miscible with alcohol, chloroform and ether | |
| OCC=C | |
| 58.08 | |
| CHEBI:16605 | |
| 99% | |
| Liquid |
Chemical Identifiers
| 107-18-6 | |
| 58.08 | |
| XXROGKLTLUQVRX-UHFFFAOYSA-N | |
| 7858 | |
| prop-2-en-1-ol |
| C3H6O | |
| MFCD00002920 | |
| allyl alcohol, 2-propen-1-ol, vinylcarbinol, 2-propenyl alcohol, 3-hydroxypropene, 2-propenol, allylic alcohol, 1-propen-3-ol, vinyl carbinol, weed drench | |
| CHEBI:16605 | |
| OCC=C |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air
GHS Signal Word: Danger
EINECSNumber : 203-470-7
RUO – Research Use Only