missing translation for 'onlineSavingsMsg'
Learn More

Allyl alcohol, 98+%

Catalog No. AAA15026AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15026AE 100 mL
AAA15026AP 500 mL
AAA150260F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA15026AE Supplier Thermo Scientific Chemicals Supplier No. A15026AE
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 107-18-6 | C3H6O | 58.08 g/mol

Allyl alcohol is widely used as a precursor to many specialized compounds such as allyl glycidyl ether, allyl diglycidyl carbonate, allyl methacrylate, flame-resistant materials, drying oils and plasticizers. It is used as a raw material for the synthesis of glycerol, glycidyl ethers, esters and amines. It is involved in the synthesis of a variety of polymerizable esters such as diallyl phthalate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl alcohol is widely used as a precursor to many specialized compounds such as allyl glycidyl ether, allyl diglycidyl carbonate, allyl methacrylate, flame-resistant materials, drying oils and plasticizers. It is used as a raw material for the synthesis of glycerol, glycidyl ethers, esters and amines. It is involved in the synthesis of a variety of polymerizable esters such as diallyl phthalate.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, acids, oleum and alkali metals. Polymerizes during prolonged storage.

Chemical Identifiers

CAS 107-18-6
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00002920
InChI Key XXROGKLTLUQVRX-UHFFFAOYSA-N
Synonym allyl alcohol, 2-propen-1-ol, vinylcarbinol, 2-propenyl alcohol, 3-hydroxypropene, 2-propenol, allylic alcohol, 1-propen-3-ol, vinyl carbinol, weed drench
PubChem CID 7858
ChEBI CHEBI:16605
IUPAC Name prop-2-en-1-ol
SMILES OCC=C

Specifications

Melting Point -129°C
Density 0.852
Boiling Point 95°C to 97°C
Flash Point 22°C (71°F)
Odor Pungent
Assay Percent Range ≥98%
Refractive Index 1.413
Linear Formula CH2=CHCH2OH
Quantity 100 mL
UN Number UN1098
Beilstein 605307
Merck Index 14,286
Solubility Information Miscible with water.
Formula Weight 58.08
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Allyl alcohol
Show More Show Less
Hazard Category H225-H301-H310+H330-H315-H319-H335
Hazard Statement GHS H Statement
H225-H301-H311-H331-H315-H319-H335
Highly flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALLYL ALCOHOL
EINECSNumber 203-470-7
RTECSNumber BA5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only