missing translation for 'onlineSavingsMsg'
Learn More

Allyl acrylate, 95%, stab. with 4-methoxyphenol, Thermo Scientific Chemicals

$718.48 - $718.48

Chemical Identifiers

CAS 999-55-3
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00014949
InChI Key QTECDUFMBMSHKR-UHFFFAOYSA-N
Synonym allyl acrylate, acrylic acid, allyl ester, 2-propenoic acid, 2-propenyl ester, unii-705ga5o3us, ccris 4745, acrylic acid allyl ester, 2-propenoic acid, 2-propen-1-yl ester, allylacrylate, acrylic acid allyl, acmc-20alut
PubChem CID 13835
IUPAC Name prop-2-enyl prop-2-enoate
SMILES C=CCOC(=O)C=C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4446314
View Documents
Thermo Scientific Chemicals
04446314
25 g N/A
Only null left
 
Description

Description

Allyl acrylate is a monofunctional monomer which function as a chain stopper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

999-55-3
112.128
QTECDUFMBMSHKR-UHFFFAOYSA-N
13835
C=CCOC(=O)C=C
C6H8O2
MFCD00014949
allyl acrylate, acrylic acid, allyl ester, 2-propenoic acid, 2-propenyl ester, unii-705ga5o3us, ccris 4745, acrylic acid allyl ester, 2-propenoic acid, 2-propen-1-yl ester, allylacrylate, acrylic acid allyl, acmc-20alut
prop-2-enyl prop-2-enoate

Specifications

999-55-3
46°C to 47°C (40 mmHg)
Pungent
1.436
25 g
1743576
Slightly soluble in water.
C=CCOC(=O)C=C
112.128
112.13
Liquid
0.935 g/mL
32°C (89°F)
C6H8O2
MFCD00014949
UN2929
allyl acrylate, acrylic acid, allyl ester, 2-propenoic acid, 2-propenyl ester, unii-705ga5o3us, ccris 4745, acrylic acid allyl ester, 2-propenoic acid, 2-propen-1-yl ester, allylacrylate, acrylic acid allyl, acmc-20alut
QTECDUFMBMSHKR-UHFFFAOYSA-N
prop-2-enyl prop-2-enoate
13835
95%
Allyl acrylate, Stabilized with 4-methoxyphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H310-H330-H226-H302-H315-H319-H335
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H311-H315-H319-H330-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

EINECSNumber : 213-662-2

RTECSNumber : UD3675000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only