missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl Acetoacetate 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | -85°C |
| Color | Yellow |
| Boiling Point | 195°C |
| Quantity | 25 g |
| UN Number | 2810 |
| Formula Weight | 142.15 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Allyl Acetoacetate |
Safety and Handling
| EINECSNumber | (2)-3495 |
| TSCA | No |