missing translation for 'onlineSavingsMsg'
Learn More

Allyl Acetate 97.0+%, TCI America™

Catalog No. p-7116661
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
A002025ML 25 mL
A0020500ML 500 mL
2 options

Catalog No. A002025ML

Supplier: TCI America A002025ML

Only null left

Chemical Identifiers

CAS 591-87-7
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00008721
InChI Key FWZUNOYOVVKUNF-UHFFFAOYSA-N
Synonym allyl acetate, 3-acetoxypropene, acetic acid, 2-propenyl ester, 2-propenyl acetate, 2-propenyl ethanoate, 3-acetoxy-1-propene, allylacetate, acetic acid, allyl ester, 2-propenyl methanoate, unii-e4u5e5990i
PubChem CID 11584
IUPAC Name prop-2-enyl acetate
SMILES CC(=O)OCC=C

Specifications

Color Colorless
Boiling Point 104°C
Quantity 25 mL
UN Number 2333
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Acetate
EINECSNumber (2)-0730&(2)-0759
RTECSNumber AF1750000
TSCA Yes
Recommended Storage Refrigerator