missing translation for 'onlineSavingsMsg'
Learn More

Allyl Acetate 97.0+%, TCI America™

Catalog No. A002025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A002025ML 25 mL
A0020500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A002025ML Supplier TCI America Supplier No. A002025ML

Chemical Identifiers

CAS 591-87-7
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00008721
InChI Key FWZUNOYOVVKUNF-UHFFFAOYSA-N
Synonym allyl acetate, 3-acetoxypropene, acetic acid, 2-propenyl ester, 2-propenyl acetate, 2-propenyl ethanoate, 3-acetoxy-1-propene, allylacetate, acetic acid, allyl ester, 2-propenyl methanoate, unii-e4u5e5990i
PubChem CID 11584
IUPAC Name prop-2-enyl acetate
SMILES CC(=O)OCC=C

Specifications

Color Colorless
Boiling Point 104°C
Quantity 25 mL
UN Number 2333
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Acetate
EINECSNumber (2)-0730&(2)-0759
RTECSNumber AF1750000
TSCA Yes
Recommended Storage Refrigerator