Learn More
Allyl acetate, 99%
CAS: 591-87-7 | C5H8O2 | 100.12 g/mol
$91.90 - $91.90
Chemical Identifiers
| CAS | 591-87-7 |
|---|---|
| MDL Number | MFCD00008721 |
| InChI Key | FWZUNOYOVVKUNF-UHFFFAOYSA-N |
| Synonym | allyl acetate, 3-acetoxypropene, acetic acid, 2-propenyl ester, 2-propenyl acetate, 2-propenyl ethanoate, 3-acetoxy-1-propene, allylacetate, acetic acid, allyl ester, 2-propenyl methanoate, unii-e4u5e5990i |
| PubChem CID | 11584 |
| IUPAC Name | prop-2-enyl acetate |
| SMILES | CC(=O)OCC=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC180651000
|
Thermo Scientific Chemicals
180651000 |
100 mL |
Each for $91.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 591-87-7 | |
| FWZUNOYOVVKUNF-UHFFFAOYSA-N | |
| 11584 | |
| CC(=O)OCC=C |
| MFCD00008721 | |
| allyl acetate, 3-acetoxypropene, acetic acid, 2-propenyl ester, 2-propenyl acetate, 2-propenyl ethanoate, 3-acetoxy-1-propene, allylacetate, acetic acid, allyl ester, 2-propenyl methanoate, unii-e4u5e5990i | |
| prop-2-enyl acetate |
Specifications
| 591-87-7 | |
| 100.0 | |
| Colorless | |
| 103.0°C to 104.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4030 to 1.4050 | |
| MFCD00008721 | |
| 02, 136 | |
| 15, 276 | |
| Solubility in water: insoluble | |
| CC(=O)OCC=C | |
| 100.12 | |
| 100.12 | |
| Liquid |
| 98.5 | |
| -96.0°C | |
| 0.9200g/mL | |
| 6°C | |
| 99% | |
| C5H8O2 | |
| CH3CO2CH2CH=CH2 | |
| 100 mL | |
| 0.92 | |
| allyl acetate, 3-acetoxypropene, acetic acid, 2-propenyl ester, 2-propenyl acetate, 2-propenyl ethanoate, 3-acetoxy-1-propene, allylacetate, acetic acid, allyl ester, 2-propenyl methanoate, unii-e4u5e5990i | |
| FWZUNOYOVVKUNF-UHFFFAOYSA-N | |
| prop-2-enyl acetate | |
| 11584 | |
| 99% | |
| Allyl acetate, 99% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed.
Harmful in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 209-734-8
RTECSNumber : AF1750000
TSCA : TSCA
RUO – Research Use Only