missing translation for 'onlineSavingsMsg'
Learn More

Allyl acetate, 97%

Catalog No. p-7051496
Change view
Click to view available options
Quantity:
50 mL
250 mL
Catalog No. Quantity
AA41353AD 50 mL
AA41353AK 250 mL
2 options

Catalog No. AA41353AD

Supplier: Thermo Scientific Chemicals 041353AD

Only null left

CAS: 591-87-7 | C5H8O2 | 100.117 g/mol

Ally acetate is used in industrial intermediate. Used in synthesis of colophony and bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 591-87-7
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00008721
InChI Key FWZUNOYOVVKUNF-UHFFFAOYSA-N
Synonym allyl acetate, 3-acetoxypropene, acetic acid, 2-propenyl ester, 2-propenyl acetate, 2-propenyl ethanoate, 3-acetoxy-1-propene, allylacetate, acetic acid, allyl ester, 2-propenyl methanoate, unii-e4u5e5990i
PubChem CID 11584
IUPAC Name prop-2-enyl acetate
SMILES CC(=O)OCC=C

Specifications

Melting Point 6°C
Density 0.928 g/mL
Boiling Point 103°C to 104°C
Refractive Index 1.4042
Quantity 50 mL
UN Number UN2333
Merck Index 14,285
Solubility Information Soluble in ethanol. Miscible with ethanol and ether. Slightly soluble in water.
Formula Weight 100.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Allyl acetate
Show More Show Less
Hazard Category H225-H301+H331-H312-H319
Hazard Statement GHS H Statement
H225-H301-H312
Highly flammable liquid and vapor.
Toxic if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALLYL ACETATE
EINECSNumber 209-734-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only