missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allura Red AC, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 496.42 |
| Chemical Name or Material | Allura Red AC |
Safety and Handling
| RTECSNumber | QK2260000 |
| TSCA | Yes |