missing translation for 'onlineSavingsMsg'
Learn More

Allura Red AC, TCI America™

Supplier:  TCI America A094325G

Quantity 25 g

 View more versions of this product

Catalog No. A094325G


Only null left

Description

Description

Specifications

Chemical Identifiers

25956-17-6
496.42
POXPUHKJGLHZEM-VIPPSAFOSA-L
44386827
[Na+].[Na+].COC1=CC(=C(C)C=C1N\N=C1\C(=O)C=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O
C18H14N2Na2O8S2
MFCD00059526
FD and C Red No.40
disodium (5E)-5-[2-(2-methoxy-5-methyl-4-sulfonatophenyl)hydrazin-1-ylidene]-6-oxo-5,6-dihydronaphthalene-2-sulfonate

Specifications

Allura Red AC
C18H14N2Na2O8S2
FD and C Red No.40
[Na+].[Na+].COC1=CC(=C(C)C=C1N\N=C1\C(=O)C=CC2=CC(=CC=C12)S([O-])(=O)=O)S([O-])(=O)=O
496.42
496.42
Crystal/Powder
25956-17-6
MFCD00059526
POXPUHKJGLHZEM-VIPPSAFOSA-L
disodium (5E)-5-[2-(2-methoxy-5-methyl-4-sulfonatophenyl)hydrazin-1-ylidene]-6-oxo-5,6-dihydronaphthalene-2-sulfonate
44386827
25 g
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : QK2260000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions