Learn More
Waters Limited Alliance HT Qualification Standards Kit
Catalog No. 50201665
Encompass
Alliance HPLC standards kit for system assessment. Kit contains standards appropriate for verification of system reproducibility, detector linearity, detector accuracy, injector accuracy,injector carryover.
Catalog No. 50201665 Supplier Waters Limited Supplier No. 700002390
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Alliance HPLC standards kit for system assessment. Kit contains standards appropriate for verification of system reproducibility, detector linearity, detector accuracy, injector accuracy,injector carryover.