missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allantoin, USP, 98.5-101%, Spectrum™ Chemical
Description
Spectrum™ Chemical Allantoin, USP promotes cell proliferation and wound healing and has a soothing, anti-irritant, and skin protectant effect. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Ignition Residue | 0.1% |
| Loss on Drying | 0.1% |
| Packaging | Poly Bottle |
| Quantity | 100 g |
| Percent Purity | 98.5 to 101% |
| Grade | USP |