missing translation for 'onlineSavingsMsg'
Learn More

Allantoin, Powder, 98%, Spectrum™ Chemical

Catalog No. p-7165104
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608479 100 g Poly Bottle
18608483 500 g Poly Bottle
18608481 2.5 kg Poly Bottle
18608480 12 kg Poly Pail
18608482 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608479 Supplier Spectrum Chemical Mfg Cor Supplier No. AL213100GM
Only null left

Chemical Identifiers

CAS 97-59-6

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 100 g
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.