missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allantoin, Powder, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Poly Bottle |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 158.12 |
| Percent Purity | 0.98 |
| Grade | Reagent |
| Physical Form | Powder or Waxy Solid |
| Chemical Name or Material | Allantoin |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.