missing translation for 'onlineSavingsMsg'
Learn More

Allantoin, 98%

Catalog No. p-51951
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC102841000 100 g
AC102845000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102841000 Supplier Thermo Scientific Chemicals Supplier No. 102841000
Only null left

CAS: 97-59-6 | C4H6N4O3 | 158.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-59-6
Molecular Formula C4H6N4O3
Molecular Weight (g/mol) 158.12
MDL Number MFCD00005260
InChI Key POJWUDADGALRAB-UHFFFAOYSA-N
Synonym allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream
PubChem CID 204
ChEBI CHEBI:15676
IUPAC Name (2,5-dioxoimidazolidin-4-yl)urea
SMILES C1(C(=O)NC(=O)N1)NC(=O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 230.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Beilstein 25, V,15, 338
Merck Index 15, 250
Solubility Information Solubility in water: 1g/190mL. Other solubilities: readily soluble in alkalies,1g/500mL alcohol,practically insoluble in ether
Formula Weight 158.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Allantoin, 98%
Show More Show Less
EINECSNumber 202-592-8
RTECSNumber YT1600000
TSCA TSCA

RUO – Research Use Only