missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals all-trans-Retinoic acid, 97%

Catalog No. AC207340010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207340010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC207340010 Supplier Thermo Scientific Chemicals Supplier No. 207340010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

All-trans-retinol, Vitamin A, retinol, CAS # 68-26-8, is a fat-soluble vitamin that binds retinoid receptors within cells. | CAS: 302-79-4 | C20H28O2 | 300.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient
• All-trans-retinol is involved in hormonal signaling via the retinoid receptors
• It is an important regulator of cell division and apoptosis
• Deficiency can lead to vision, bone, immune, and skin disorders

Applications
• Suitable for research on cell differentiation, embryonic development, immunology and more
• Laboratory grade quality, for research use only

Chemical Identifiers

CAS 302-79-4
Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.44
MDL Number MFCD00001551
InChI Key SHGAZHPCJJPHSC-YCNIQYBTSA-N
Synonym retinoic acid, tretinoin, vitamin a acid, all-trans-retinoic acid, trans-retinoic acid, renova, airol, retin-a, vesanoid, atra
PubChem CID 444795
ChEBI CHEBI:15367
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
SMILES C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 179°C to 184°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 09, III, 3118
Merck Index 15, 8288
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,dmso (25mg/mL),slightly soluble in alcohol and chloroform
Formula Weight 300.44
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material all-trans-Retinoic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-129-0
RTECSNumber VH6475000
TSCA TSCA

RUO – Research Use Only